ENAH38A2CFAF Enamine

3-bromo-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-2-carboxylic acid hydrobromide

  • MDL number MFCD31810789
  • Cas Number : 2305253-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762243
Availability details for
Quantity
  • Availability
  • Add to Cart