ENAH38A2CFB5 Enamine

tert-butyl 3-[5-amino-2-(methoxycarbonyl)phenoxy]pyrrolidine-1-carboxylate

  • MDL number MFCD30990117
  • Cas Number : 2309448-63-1
  • Molecular Weight : 336.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762297
Availability details for
Quantity
  • Availability
  • Add to Cart