ENAH38A2CFBF Enamine

tert-butyl N-{2-[4-(chloromethyl)-1,3-oxazol-5-yl]ethyl}carbamate

  • MDL number MFCD31810797
  • Cas Number : 2305255-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762332
Availability details for
Quantity
  • Availability
  • Add to Cart