ENAH38A2CFC8 Enamine

N'-hydroxy-6-(4-methyl-1,4-diazepan-1-yl)pyridine-3-carboximidamide hydrochloride

  • Cas Number : 2305263-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762356
Availability details for
Quantity
  • Availability
  • Add to Cart