ENAH38A2CFCB Enamine

4,4-difluoro-2,2-dimethylcyclohexan-1-amine hydrochloride

  • MDL number MFCD31810566
  • Cas Number : 2305255-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762382
Availability details for
Quantity
  • Availability
  • Add to Cart