ENAH38A2CFCC Enamine

2-[2-(4-fluorophenyl)-5-methyl-1,3-oxazol-4-yl]acetic acid

  • MDL number MFCD07378988
  • Cas Number : 407640-15-7
  • Molecular Weight : 235.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762384
Availability details for
Quantity
  • Availability
  • Add to Cart