ENAH38A2CFDC Enamine

7-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD31810568
  • Cas Number : 1545012-38-1
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762509
Availability details for
Quantity
  • Availability
  • Add to Cart