ENAH38A2CFDF Enamine

2-oxo-1,3-dioxaindane-5-carboxylic acid

  • MDL number MFCD20640629
  • Cas Number : 861605-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762526
Availability details for
Quantity
  • Availability
  • Add to Cart