ENAH38A2CFF1 Enamine

diethyl [(S)-amino(phenyl)methyl]phosphonate hydrochloride

  • MDL number MFCD26547857
  • Cas Number : 59005-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762579
Availability details for
Quantity
  • Availability
  • Add to Cart