ENAH38A2CFF4 Enamine

1-(4-aminophenyl)-3-cyclopentylurea

  • MDL number MFCD12176738
  • Cas Number : 1153357-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762593
Availability details for
Quantity
  • Availability
  • Add to Cart