ENAH38A2D7FC Enamine

5-methoxybicyclo[2.2.1]heptan-2-amine hydrochloride

  • MDL number MFCD31810599
  • Cas Number : 2305252-40-6
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762660
Availability details for
Quantity
  • Availability
  • Add to Cart