ENAH38A2D800 Enamine

methyl (2S,4S)-4-chloropyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD31810602
  • Cas Number : 2305184-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762771
Availability details for
Quantity
  • Availability
  • Add to Cart