ENAH38A2D805 Enamine

(2S)-1-(trifluoromethoxy)butan-2-amine hydrochloride

  • MDL number MFCD31810606
  • Cas Number : 2305202-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762896
Availability details for
Quantity
  • Availability
  • Add to Cart