ENAH38A2D807 Enamine

5-(bromomethyl)-1-methyl-2-oxabicyclo[3.1.1]heptane

  • MDL number MFCD31810607
  • Cas Number : 2305252-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762922
Availability details for
Quantity
  • Availability
  • Add to Cart