ENAH38A2D811 Enamine

2-[(2S)-2,3-dihydro-1H-indol-2-yl]acetic acid hydrochloride

  • MDL number MFCD31810974
  • Cas Number : 2305185-14-0
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763312
Availability details for
Quantity
  • Availability
  • Add to Cart