ENAH38A2D814 Enamine

2-{6-oxaspiro[3.4]octan-7-yl}ethane-1-sulfonyl chloride

  • MDL number MFCD31810982
  • Cas Number : 2305254-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763410
Availability details for
Quantity
  • Availability
  • Add to Cart