ENAH38A2D815 Enamine

4-methyl-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD31810649
  • Cas Number : 2305251-68-5
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763412
Availability details for
Quantity
  • Availability
  • Add to Cart