ENAH38A2D816 Enamine

3-oxaspiro[bicyclo[3.1.0]hexane-2,3'-pyrrolidine] hydrochloride

  • MDL number MFCD31810650
  • Cas Number : 2305251-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763453
Availability details for
Quantity
  • Availability
  • Add to Cart