ENAH38A2D818 Enamine

3-amino-2-(prop-2-en-1-yl)cyclobutan-1-ol

  • MDL number MFCD31810986
  • Cas Number : 2305255-56-3
  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763463
Availability details for
Quantity
  • Availability
  • Add to Cart