ENAH38A2D81A Enamine

5-methoxypiperidin-3-ol hydrochloride

  • MDL number MFCD31810655
  • Cas Number : 2305253-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763507
Availability details for
Quantity
  • Availability
  • Add to Cart