ENAH38A2D81D Enamine

lithium(1+) ion 2-tert-butyl-1,3-oxazole-5-sulfinate

  • MDL number MFCD31811042
  • Cas Number : 2305254-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763536
Availability details for
Quantity
  • Availability
  • Add to Cart