ENAH38A2D820 Enamine

[3-(1-methyl-1H-pyrazol-5-yl)-1H-pyrazol-4-yl]methanamine hydrochloride

  • MDL number MFCD31915252
  • Cas Number : 2309460-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352575
Availability details for
Quantity
  • Availability
  • Add to Cart