ENAH38A2D821 Enamine

3-(chloromethyl)-4-methyl-5-phenyl-1H-pyrazole hydrochloride

  • MDL number MFCD31915277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352739
Availability details for
Quantity
  • Availability
  • Add to Cart