ENAH38A2D825 Enamine

4-(dimethylphosphanyl)pyridine

  • MDL number MFCD31810600
  • Cas Number : 2305254-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762682
Availability details for
Quantity
  • Availability
  • Add to Cart