ENAH38A2D828 Enamine

2-phenyl-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine dihydrochloride

  • MDL number MFCD29762534
  • Cas Number : 485402-40-2
  • Molecular Weight : 272.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762932
Availability details for
Quantity
  • Availability
  • Add to Cart