ENAH38A2D82D Enamine

methyl 2-(1H-1,2,3-triazol-1-yl)acetate

  • MDL number MFCD22548215
  • Cas Number : 1021869-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763013
Availability details for
Quantity
  • Availability
  • Add to Cart