ENAH38A2D83A Enamine

2-chloro-3-(chloromethyl)-6-methoxyquinoline

  • MDL number MFCD09787660
  • Cas Number : 948291-11-0
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763053
Availability details for
Quantity
  • Availability
  • Add to Cart