ENAH38A2D83E Enamine

6-oxa-7lambda6-thia-8-azaspiro[3.5]nonane-7,7-dione

  • MDL number MFCD31810637
  • Cas Number : 2305252-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763060
Availability details for
Quantity
  • Availability
  • Add to Cart