ENAH38A2D83F Enamine

6,6-dimethyl-1H,2H,3H,4H,5H,6H-pyrrolo[3,4-c]pyrazol-3-one dihydrochloride

  • MDL number MFCD31810638
  • Cas Number : 2305252-99-5
  • Molecular Weight : 226.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763061
Availability details for
Quantity
  • Availability
  • Add to Cart