ENAH38A2D840 Enamine

3-(1-methyl-1H-1,3-benzodiazol-5-yl)-3-oxopropanenitrile

  • MDL number MFCD31810639
  • Cas Number : 1616099-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763064
Availability details for
Quantity
  • Availability
  • Add to Cart