ENAH38A2D843 Enamine

methyl 5-(1,1-difluoroethyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD31915504
  • Cas Number : 2309450-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763244
Availability details for
Quantity
  • Availability
  • Add to Cart