ENAH38A2D847 Enamine

3-bromotricyclo[2.2.1.0,2,6]heptane

  • Cas Number : 695-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763284
Availability details for
Quantity
  • Availability
  • Add to Cart