ENAH38A2D852 Enamine

(but-3-en-1-yl)(pentyl)amine

  • MDL number MFCD20289040
  • Cas Number : 1339032-03-9
  • Molecular Weight : 141.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763360

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6763360
pictograms GHS05,GHS07
hstatements H227,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart