ENAH38A2D858 Enamine

2-methoxy-1-methyl-3-(pyrrolidin-2-yl)-1H-indole

  • MDL number MFCD31915484
  • Cas Number : 2309449-80-5
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762793
Availability details for
Quantity
  • Availability
  • Add to Cart