ENAH38A2D85F Enamine

(3S)-hexan-3-ol

  • MDL number MFCD30076489
  • Cas Number : 6210-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762808
Availability details for
Quantity
  • Availability
  • Add to Cart