ENAH38A2D861 Enamine

(2S,4S)-1-[(tert-butoxy)carbonyl]-4-chloropyrrolidine-2-carboxylic acid

  • MDL number MFCD31810983
  • Cas Number : 183108-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763414
Availability details for
Quantity
  • Availability
  • Add to Cart