ENAH38A2D864 Enamine

(2R)-1-methoxy-3-(2-nitro-1H-imidazol-1-yl)propan-2-ol

  • MDL number MFCD31915507
  • Cas Number : 95120-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763433
Availability details for
Quantity
  • Availability
  • Add to Cart