ENAH38A2D86A Enamine

1-[(2-hydroxy-3,3-dimethylbutyl)(methyl)amino]-3,3-dimethylbutan-2-ol

  • MDL number MFCD31810652
  • Cas Number : 2305253-08-9
  • Molecular Weight : 231.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763466
Availability details for
Quantity
  • Availability
  • Add to Cart