ENAH38A2D86B Enamine

5,7-dimethyl-1,4-diazepane

  • MDL number MFCD19217449
  • Cas Number : 16698-08-1
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763467
Availability details for
Quantity
  • Availability
  • Add to Cart