ENAH38A2D873 Enamine

6-(4-fluorophenyl)-4-oxo-4H,5H-[1,2,3]triazolo[1,5-a]pyrazine-3-carboxylic acid

  • MDL number MFCD28963007
  • Cas Number : 2108267-47-4
  • Molecular Weight : 274.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763501
Availability details for
Quantity
  • Availability
  • Add to Cart