ENAH38A2D87A Enamine

{8-oxa-1-azaspiro[4.5]decan-3-yl}methanol

  • MDL number MFCD31810866
  • Cas Number : 2305255-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6762650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6762650
Availability details for
Quantity
  • Availability
  • Add to Cart