ENAH38A2D87F Enamine

methyl 4-iodo-3-methyl-1,2-oxazole-5-carboxylate

  • MDL number MFCD31915831
  • Cas Number : 2309457-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763591
Availability details for
Quantity
  • Availability
  • Add to Cart