ENAH38A2D880 Enamine

(ethylsulfanyl)carbothioamide

  • MDL number MFCD19203422
  • Cas Number : 625-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763631
Availability details for
Quantity
  • Availability
  • Add to Cart