ENAH38A2D889 Enamine

7-fluoro-6-methoxyquinoline

  • MDL number MFCD19690247
  • Cas Number : 887769-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764020
Availability details for
Quantity
  • Availability
  • Add to Cart