ENAH38A2D893 Enamine

5-methyl-4,5-dihydro-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD19228939
  • Cas Number : 4486-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764115
Availability details for
Quantity
  • Availability
  • Add to Cart