ENAH38A2D894 Enamine

2-chloro-2,2-difluoroethanecarbonyl isothiocyanate

  • MDL number MFCD31915518
  • Cas Number : 1446015-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -70
Physical Form liquid
Shipping Temperature Dry Ice
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764126
Availability details for
Quantity
  • Availability
  • Add to Cart