ENAH38A2D89B Enamine

potassium {bicyclo[4.1.0]heptan-1-yl}trifluoroboranuide

  • MDL number MFCD31915522
  • Molecular Weight : 202.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764147
Availability details for
Quantity
  • Availability
  • Add to Cart