ENAH38A2D8A3 Enamine

2-cyclopentylpropane-1-sulfonyl chloride

  • MDL number MFCD31915842
  • Cas Number : 1889994-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764162
Availability details for
Quantity
  • Availability
  • Add to Cart