ENAH38A2D8A4 Enamine

(2R)-1-(2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-6-yl)propan-2-amine hydrochloride

  • MDL number MFCD31915527
  • Cas Number : 2309431-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764165
Availability details for
Quantity
  • Availability
  • Add to Cart