ENAH38A2D8A6 Enamine

1-[5-(1-aminoethyl)-4H-1,2,4-triazol-3-yl]ethan-1-ol hydrochloride

  • MDL number MFCD31915528
  • Cas Number : 2309468-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6764170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764170
Availability details for
Quantity
  • Availability
  • Add to Cart